missing translation for 'onlineSavingsMsg'
Learn More

Sodium metabisulfite, 97+%, ACS reagent

CAS: 7681-57-4 | Na2O5S2 | 190.09 g/mol

Supplier:  Thermo Scientific Chemicals 419580050

 View more versions of this product

Catalog No. AC419580050

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Specifications

Sodium metabisulfite
7681-57-4
Fine Crystalline Powder
97% min. (Iodimetry)
Na2S2O5
15, 8770
Solubility in water: 540g/L (20°C). Other solubilities: freely soluble in glycerol, slightly soluble in alcohol
[Na+].[Na+].[O-]S(=O)OS([O-])=O
190.09
190.1
ACS Reagent
150.0°C
White
5 g
Na2O5S2
MFCD00167602
sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
LDTLADDKFLAYJA-UHFFFAOYSA-L
disodium (sulfinatooxy)sulfinate
656671
≥97%
Glass Bottle

Chemical Identifiers

7681-57-4
190.09
LDTLADDKFLAYJA-UHFFFAOYSA-L
656671
[Na+].[Na+].[O-]S(=O)OS([O-])=O
Na2O5S2
MFCD00167602
sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
disodium (sulfinatooxy)sulfinate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Contact with acids liberates toxic gas.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

EINECSNumber : 231-673-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only