missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium metabisulfite, 97+%, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Specifications
| Melting Point | 150.0°C |
| Color | White |
| Physical Form | Fine Crystalline Powder |
| Quantity | 250 g |
| Packaging | Plastic bottle |
| Assay Percent Range | 97% min. (Iodimetry) |
| Linear Formula | Na2S2O5 |
| Merck Index | 15, 8770 |
| Solubility Information | Solubility in water: 540g/L (20°C). Other solubilities: freely soluble in glycerol,slightly soluble in alcohol |
| Formula Weight | 190.1 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes serious eye damage. Contact with acids liberates toxic gas. |
| Precautionary Statement | GHS P Statement Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physici |
| EINECSNumber | 231-673- |
RUO – Research Use Only