missing translation for 'onlineSavingsMsg'
Learn More

Sodium Meclofenamate 98.0+%, TCI America™

Catalog No. M126910G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M126910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M126910G Supplier TCI America Supplier No. M126910G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6385-02-0
Molecular Formula C14H13Cl2NNaO2
Molecular Weight (g/mol) 321.153
MDL Number MFCD00077376
InChI Key JKTYCTJLWZYQGM-UHFFFAOYSA-N
Synonym meclofenamate sodium
PubChem CID 131673979
IUPAC Name 2-(2,6-dichloro-3-methylanilino)benzoic acid;molecular hydrogen;sodium
SMILES [HH].CC1=C(C(=C(C=C1)Cl)NC2=CC=CC=C2C(=O)O)Cl.[Na]

Specifications

Melting Point 287°C
Color White
Quantity 10 g
Formula Weight 318.13
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Meclofenamate
RTECSNumber CB2975500
TSCA No