missing translation for 'onlineSavingsMsg'
Learn More

Sodium Linoleate 95.0+%, TCI America™

Catalog No. p-7131798
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L00565G 5 g
L005625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L00565G Supplier TCI America Supplier No. L00565G
Only null left

Chemical Identifiers

CAS 822-17-3
Molecular Formula C18H31NaO2
Molecular Weight (g/mol) 302.434
MDL Number MFCD00063202
InChI Key WYPBVHPKMJYUEO-UHFFFAOYSA-M
Synonym Linoleic Acid Sodium Salt
PubChem CID 44134802
IUPAC Name sodium;octadeca-9,12-dienoate
SMILES CCCCCC=CCC=CCCCCCCCC(=O)[O-].[Na+]

Specifications

Melting Point 192°C
Color White-Yellow
Quantity 5 g
Formula Weight 302.43
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Linoleate
EINECSNumber (2)-0611&(7)-0973
TSCA Yes
Recommended Storage Refrigerator