missing translation for 'onlineSavingsMsg'
Learn More

Sodium Ligninsulfonate, TCI America™

This product is chemically-processing lignin derived from natural sources.

$67.11 - $150.98

Specifications

Color Deep Reddish Yellow-deep Yellow Red
Content And Storage 20°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L0098500G
View Documents
TCI America
L0098500G
500g N/A
Only null left
 
L009825G
View Documents
TCI America
L009825G
25g N/A
Only null left
 
Description

Description

Specifications

Specifications

Deep Reddish Yellow-deep Yellow Red
20°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'