missing translation for 'onlineSavingsMsg'
Learn More

Sodium L-(+)-Tartrate Dihydrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. p-192917
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S433100 100 g Glass Bottle
S433500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S433100 Supplier Thermo Fisher Scientific Supplier No. S433100
Only null left

Chemical Identifiers

CAS 6106-24-7
Molecular Formula C4H8Na2O8
Molecular Weight (g/mol) 230.08
MDL Number MFCD00150035
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
Synonym unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi
PubChem CID 131855972
IUPAC Name disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

Melting Point 150°C
Color White
Density 1.8g/mL
pH 7 to 9
Loss on Drying 14 to 17%
Assay Percent Range ≥99.0 %
Packaging Glass Bottle
Quantity 100 g
Formula Weight 230.08
Percent Purity ≥99.0%
Grade Certified
Physical Form Powder or Solid
Chemical Name or Material Sodium L-(+)-Tartrate Dihydrate
Show More Show Less
Hazard Category SIGNAL WORD: CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation.
NFPA
Health: 1
Flammability: 1
Instability: 0