missing translation for 'onlineSavingsMsg'
Learn More

Sodium L-lactate, 98+%

Catalog No. AAL1450014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1450014 25 g
AAL1450006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1450014 Supplier Thermo Scientific Chemicals Supplier No. L1450014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 867-56-1 ∣ C3H5NaO3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications

Sodium L-lactate is used as a food additive, preservative, acidity regulator and bulking agent. It finds application in shampoo and liquid soaps and other related products. It acts as an effective humectant and moisturizer. It is used in the treatment of arrhythmias.

Solubility

Soluble in water.

Notes

Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 867-56-1
Molecular Formula C3H5NaO3
MDL Number MFCD00066576
Synonym sodium l-lactate, sodium-l-lactate, sodium lactate, l, sodium s-2-hydroxypropanoate, unii-p2y1c6m9ps, sodium s-lactate, sodium l-+-lactate, p2y1c6m9ps, propanoic acid, 2-hydroxy-, monosodium salt, 2s, l-+-lactic acid sodium salt

Specifications

Melting Point ∼160°C
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 25 g
Sensitivity Hygroscopic
Beilstein 4567087
Solubility Information Soluble in water.
Molecular Weight (g/mol) 112.06
Formula Weight 112.06
Percent Purity ≥98%
Grade Ungraded
Chemical Name or Material Sodium L-lactate
Show More Show Less
EINECSNumber 212-762-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only