missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium L-Glutamate Monohydrate 99.0+%, TCI America™
$34.66 - $50.05
Chemical Identifiers
| CAS | 6106-04-3 |
|---|---|
| Molecular Formula | C5H10NNaO5 |
| Molecular Weight (g/mol) | 187.13 |
| MDL Number | MFCD00013074 |
| InChI Key | GJBHGUUFMNITCI-QTNFYWBSSA-M |
| Synonym | monosodium glutamate, l-glutamic acid monosodium salt monohydrate, p.a, l-glutamic acid monosodium salt monohydrate nt, l-glutamic acid monosodium salt monohydrate, saj special grade, monosodium glutamate, united states pharmacopeia usp reference standard, l-glutamic acid, monosodium salt, low endotoxin, pharmagrade, manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production., suitable for cell culture |
| PubChem CID | 87090819 |
| IUPAC Name | (2S)-2-aminopentanedioic acid;sodium;hydrate |
| SMILES | C(CC(=O)O)C(C(=O)O)N.O.[Na] |
Chemical Identifiers
| 6106-04-3 | |
| 187.13 | |
| GJBHGUUFMNITCI-QTNFYWBSSA-M | |
| 87090819 | |
| C(CC(=O)O)C(C(=O)O)N.O.[Na] |
| C5H10NNaO5 | |
| MFCD00013074 | |
| monosodium glutamate, l-glutamic acid monosodium salt monohydrate, p.a, l-glutamic acid monosodium salt monohydrate nt, l-glutamic acid monosodium salt monohydrate, saj special grade, monosodium glutamate, united states pharmacopeia usp reference standard, l-glutamic acid, monosodium salt, low endotoxin, pharmagrade, manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production., suitable for cell culture | |
| (2S)-2-aminopentanedioic acid;sodium;hydrate |
Specifications
| 6106-04-3 | |
| C5H10NNaO5 | |
| monosodium glutamate, l-glutamic acid monosodium salt monohydrate, p.a, l-glutamic acid monosodium salt monohydrate nt, l-glutamic acid monosodium salt monohydrate, saj special grade, monosodium glutamate, united states pharmacopeia usp reference standard, l-glutamic acid, monosodium salt, low endotoxin, pharmagrade, manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production., suitable for cell culture | |
| C(CC(=O)O)C(C(=O)O)N.O.[Na] | |
| 25 g | |
| 87090819 | |
| ≥99.0% (T) | |
| Sodium L-Glutamate Monohydrate |
| White | |
| MFCD00013074 | |
| GJBHGUUFMNITCI-QTNFYWBSSA-M | |
| (2S)-2-aminopentanedioic acid;sodium;hydrate | |
| 187.13 | |
| 169.11 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-1540
RTECSNumber : MA1575000
TSCA : Yes