missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium L-Glutamate Monohydrate 99.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 25 g |
| Formula Weight | 169.11 |
| Percent Purity | ≥99.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Sodium L-Glutamate Monohydrate |
Safety and Handling
| EINECSNumber | (9)-1540 |
| RTECSNumber | MA1575000 |
| TSCA | Yes |