missing translation for 'onlineSavingsMsg'
Learn More

Sodium L-Ascorbate 98.0+%, TCI America™

Catalog No. A0539500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0539500G 500 g
A053925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0539500G Supplier TCI America Supplier No. A0539500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 134-03-2 | Formula: C6H7NaO6 | Weight: 198.11 g/mol

Chemical Identifiers

CAS 134-03-2
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
MDL Number MFCD00082340
InChI Key IFVCRSPJFHGFCG-HXPAKLQESA-N
Synonym sodium ascorbate, l-ascorbic acid sodium salt, sodium l-ascorbate, vitamin c sodium, ascorbicin, sodascorbate, cebitate, aminofenitrooxon, natrii ascorbas, monosodium l-ascorbate
PubChem CID 131674100
IUPAC Name sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
SMILES [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

Specifications

Melting Point 218°C
Color White
Physical Form Crystalline Powder
Quantity 500 g
Formula Weight 198.11
Percent Purity ≥98.0% (T)
Chemical Name or Material Sodium L-Ascorbate
RTECSNumber CI7671000
TSCA Yes