missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium, Ion chromatography standard solution, Specpure™ Na∣+ 1000μg/mL
CAS: 7440-23-5 | Na | 22.99 g/mol
Supplier: Thermo Scientific Chemicals 041696AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Sodium, Ion chromatography standard solution | |
| Plastic bottle | |
| MFCD00085307 | |
| natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, monosodium | |
| [Na] | |
| 22.99 | |
| CHEBI:26708 |
| 7440-23-5 | |
| Na | |
| 100 mL | |
| KEAYESYHFKHZAL-UHFFFAOYSA-N | |
| sodium | |
| 5360545 | |
| Liquid |
Chemical Identifiers
| 7440-23-5 | |
| 22.99 | |
| KEAYESYHFKHZAL-UHFFFAOYSA-N | |
| 5360545 | |
| sodium |
| Na | |
| MFCD00085307 | |
| natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, monosodium | |
| CHEBI:26708 | |
| [Na] |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only