missing translation for 'onlineSavingsMsg'
Learn More

Sodium iodide, Honeywell Fluka™

Catalog No. p-7072196
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001494 500 g
6001493 2.5 kg
6001492 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6001494 Supplier Honeywell Chemicals Supplier No. 383112500G
Only null left

ACS Reagent,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.894
MDL Number MFCD00003532
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
Synonym sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]

Specifications

Quantity 500 g
Linear Formula NaI
UN Number UN3077
Formula Weight 149.89g/mol
Percent Purity ≥99.5%
Density Vapor Density: >1 (vs air)
Chemical Name or Material Sodium iodide
Hazard Statement P273-P305 + P351 + P338
RTECSNumber WB6475000