missing translation for 'onlineSavingsMsg'
Learn More

Sodium Iodide (Crystalline/Certified), Fisher Chemical™

Catalog No. S324100
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S324100 100 g Glass Bottle
S324500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S324100 Supplier Thermo Fisher Scientific Supplier No. S324 100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.894
MDL Number MFCD00003532
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
Synonym sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]

Specifications

Boiling Point 1300°C
Melting Point 661°C
Color White
pH 6 to 9
Physical Form Powder/Solid
Quantity 100 g
Formula Weight 149.89
Grade Certified
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure 1.3mbar at 767°C
Density 3.660g/cm³
Chemical Name or Material Sodium Iodide
Show More Show Less