missing translation for 'onlineSavingsMsg'
Learn More

Sodium Iodide, ACS Reagent, Reagents

Catalog No. 01337718
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337718 125 g
01337716 25 g
01337715 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337718 Supplier Labchem Inc Supplier No. C1320000125D2
Only null left

Sodium Iodide, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.89
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
IUPAC Name sodium iodide
SMILES [Na+].[I-]

Specifications

Boiling Point 1304°C
Color White
Physical Form Solid
Quantity 125 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 3.67
Chemical Name or Material Sodium Iodide, ACS Reagent