missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Iodide, ACS Grade, ≥ 99.5%, LabChem™
Supplier: LabChem LC246451
Specifications
| 1,304°C | |
| 651°C | |
| White | |
| Crystals | |
| INa | |
| sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech | |
| FVAUCKIRQBBSSJ-UHFFFAOYSA-M | |
| sodium;iodide | |
| 5238 | |
| 149.89 | |
| ACS | |
| Amber Glass | |
| Iodine vapor |
| Sodium Iodide | |
| 7681-82-5 | |
| 100 | |
| 500 g | |
| NaI | |
| Soluble in water | |
| [Na+].[I-] | |
| 149.894 | |
| CHEBI:33167 | |
| ≥99.5% | |
| Passes Test | |
| 3.667g/cm3 | |
| 3.667g/cm3 |
Chemical Identifiers
| 7681-82-5 | |
| 149.894 | |
| sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech | |
| CHEBI:33167 | |
| [Na+].[I-] |
| INa | |
| FVAUCKIRQBBSSJ-UHFFFAOYSA-M | |
| 5238 | |
| sodium;iodide |
Safety and Handling
GHS H Statement
Very toxic to aquatic life.
GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Collect spillage.
Warning
EINECSNumber : 231-668-3
Recommended Storage : Room Temperature; Protect from light