missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Iodide, ≥99%, (ACS Reagent Grade), MP Biomedicals
$669.85 - $669.85
Chemical Identifiers
| CAS | 7681-82-5 |
|---|---|
| Molecular Formula | INa |
| Molecular Weight (g/mol) | 149.894 |
| InChI Key | FVAUCKIRQBBSSJ-UHFFFAOYSA-M |
| Synonym | sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech |
| PubChem CID | 5238 |
| ChEBI | CHEBI:33167 |
| IUPAC Name | sodium;iodide |
| SMILES | [Na+].[I-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19397990
|
MP Biomedicals Inc
0219397990 |
500 g |
Each for $669.85
|
|
|||||
Chemical Identifiers
| 7681-82-5 | |
| 149.894 | |
| sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech | |
| CHEBI:33167 | |
| [Na+].[I-] |
| INa | |
| FVAUCKIRQBBSSJ-UHFFFAOYSA-M | |
| 5238 | |
| sodium;iodide |
Specifications
| 7681-82-5 | |
| ≥99% | |
| sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech | |
| [Na+].[I-] | |
| 149.894 | |
| CHEBI:33167 | |
| ACS Reagent |
| 500 g | |
| INa | |
| FVAUCKIRQBBSSJ-UHFFFAOYSA-M | |
| sodium;iodide | |
| 5238 | |
| 149.9 | |
| Sodium Iodide |
Safety and Handling
Recommended Storage : Room Temperature