missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite, Sterile, Made with USP Purified Water, 5% Solution, BAKER™, J.T. Baker™

Catalog No. 02003949
Change view
Click to view available options
Quantity:
1 gL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02003949 1 gL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02003949 Supplier Avantor Supplier No. P00503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 7681-52-9
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.439
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Physical Form Liquid
Quantity 1 gL
Formula Weight 74.44
Percent Purity 5%
Grade Pure
Specific Gravity 1L = 1.11kg
Chemical Name or Material Sodium Hypochlorite, Sterile, Made with USP Purified Water