missing translation for 'onlineSavingsMsg'
Learn More

Sodium hypochlorite solution, Reagent grade, Available chlorine 10 to 15%, Solstice

Catalog No. 6001490
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001490 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001490 Supplier Solstice Supplier No. 425044250ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent Grade, available chlorine 10-15 %

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 7681-52-9
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.439
MDL Number MFCD00011120
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Physical Form Liquid
Quantity 250 mL
Linear Formula NaClO
UN Number UN1791
Formula Weight 74.44g/mol
Percent Purity 32% to 35%
Grade Reagent
Concentration or Composition (by Analyte or Components) Available chlorine 10 to 15%
Density 1.206g/mL at 25°C
Chemical Name or Material Sodium hypochlorite solution
Hazard Statement P273-P280-P305 + P351 + P338-P310