missing translation for 'onlineSavingsMsg'
Learn More

Sodium hypochlorite solution, purum, ≈10% (RT), Honeywell Fluka™

Catalog No. p-7072265
Change view
Click to view available options
Quantity:
2.5 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001488 2.5 L
6001487 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001488 Supplier Honeywell Chemicals Supplier No. 716962.5L
Only null left

Purum, ∼10% (RT)

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 7681-52-9
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.439
MDL Number MFCD00011120
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Quantity 2.5 L
Linear Formula NaClO
UN Number UN1791
Formula Weight 74.44g/mol
Percent Purity 10% (RT)
Grade purum
Concentration or Composition (by Analyte or Components) ≈10% (RT)
Density 1.21-1.26g/mL at 20°C
Chemical Name or Material Sodium hypochlorite solution
Hazard Statement P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P391