missing translation for 'onlineSavingsMsg'
Learn More

Sodium hypochlorite solution, 6-14% active chlorine basis, Honeywell Fluka™

Catalog No. p-7122046
Change view
Click to view available options
Quantity:
500 mL
6 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001486 500 mL
60044803 6 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001486 Supplier Honeywell Chemicals Supplier No. 13440500ML
Only null left

6-14% active chlorine basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7681-52-9
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.439
MDL Number MFCD00011120
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Quantity 500 mL
Linear Formula NaClO
UN Number 1791
Formula Weight 74.44g/mol
Concentration or Composition (by Analyte or Components) 6 to 14% active chlorine basis
Packaging Poly bottle
Chemical Name or Material Sodium Hypochlorite Solution
Hazard Statement P273-P280-P305 + P351 + P338-P310
Precautionary Statement H314-H400