missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite Solution, 5% Available Chlorine, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02003948
Change view
Click to view available options
Quantity:
500 mL
Packaging:
HDPE Pressure Bottom Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003948 500 mL HDPE Pressure Bottom Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02003948 Supplier Avantor Supplier No. 941601

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use

Chemical Identifiers

CAS 7681-52-9
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.439
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Physical Form Liquid
Quantity 500 mL
Formula Weight 74.44
Percent Purity 5% (Available Chlorine)
Grade BAKER ANALYZED™ Reagent
Packaging HDPE Pressure Bottom Poly Bottle
Specific Gravity 1L = 1.11kg
Chemical Name or Material Sodium Hypochlorite Solution