missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite Pentahydrate, TCI America™

Catalog No. s093925g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S093925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S093925G Supplier TCI America Supplier No. S093925G
Only null left

Chemical Identifiers

CAS 10022-70-5
Molecular Formula ClH10NaO6
Molecular Weight (g/mol) 164.514
MDL Number MFCD01941504
InChI Key FHKLOBNGYGFRSF-UHFFFAOYSA-N
PubChem CID 23700086
IUPAC Name sodium;hypochlorite;pentahydrate
SMILES O.O.O.O.O.[O-]Cl.[Na+]

Specifications

Color Yellow
Physical Form Crystalline Powder
Quantity 25 g
UN Number 3212
Formula Weight 74.44
Chemical Name or Material Sodium Hypochlorite Pentahydrate
EINECSNumber (1)-0237
RTECSNumber NH3486800
TSCA No
Recommended Storage Freezer