missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite, Certified, 2.0% to 3.0% (w/v), LabChem™

Supplier:  LabChem LC246402

 View more versions of this product

Catalog No. LC246402


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Hypochlorite
Yellow
Liquid
ClNaO
sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
SUKJFIGYRHOWBL-UHFFFAOYSA-N
sodium;hypochlorite
23665760
74.44
Certified
Poly Bottle
7681-52-9,7732-18-5
97.7,2.3
1 L
NaOCl
Soluble in water
[O-]Cl.[Na+]
74.439
CHEBI:32146
2.0 to 3.0% (w/v)
Passes Test
Hydrogen chloride; Chlorine; Phosgene

Chemical Identifiers

7681-52-9
74.439
sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
CHEBI:32146
[O-]Cl.[Na+]
ClNaO
SUKJFIGYRHOWBL-UHFFFAOYSA-N
23665760
sodium;hypochlorite
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Toxic to aquatic life.

GHS P Statement
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

EINECSNumber : 231-668-3

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions