missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite, 5% Available Chlorine, Solution, ACS, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S16694LT

 View more versions of this product

Catalog No. 18613677


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7681-52-9,7732-18-5
4 L
SUKJFIGYRHOWBL-UHFFFAOYSA-N
sodium hypochlorite
5%
1
ClNaO
[Na+].[O-]Cl
74.44
Poly Bottle

Chemical Identifiers

7681-52-9
74.44
sodium hypochlorite
ClNaO
SUKJFIGYRHOWBL-UHFFFAOYSA-N
[Na+].[O-]Cl
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.