missing translation for 'onlineSavingsMsg'
Learn More

Sodium hypochlorite, 11-15% available chlorine

Catalog No. p-4407903
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3336936 500 g
AA33369A3 2 kg
AA33369A9 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3336936 Supplier Thermo Scientific Chemicals Supplier No. 03336936

CAS: 7681-52-9 | ClNaO | 74.439 g/mol

Sodium hypochlorite is used as a disinfectant and a bleaching agent. It is used as a destainer involved in removing of mold stains, dental stains arises by fluorosis. It finds application in laundry detergents. Its aqueous solution is used as household bleach for sanitizing smooth surfaces. In a water treatment plant, it is utilized for treating dilute cyanide waste water like electroplating wastes. It is an oxidizing agent used with phase transfer catalyst to convert alcohols into corresponding carbonyl compounds. It is an important reactant in Hofmann rearrangement to prepare primary amine from primary amide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7681-52-9
Molecular Formula ClNaO
MDL Number MFCD00011120
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146

Specifications

Color Yellow
Physical Form Liquid
Quantity 500 g
Solubility Information Miscible with water.
SMILES [O-]Cl.[Na+]
IUPAC Name sodium;hypochlorite
Molecular Weight (g/mol) 74.439
Formula Weight 74.44
Percent Purity 14.5% (available chlorine)
Appearance Light yellow liquid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Density 1.2 g/cm3 at 20°C
Chemical Name or Material Sodium hypochlorite, 14.5% available chlorine
Show More Show Less
Hazard Category H290-H314-H335
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation DOT Class: 8; Packing Group: III
EINECSNumber 231-668-3
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only