missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide Solution, 1 M, For HPCE, MilliporeSigma™ Supelco™

Catalog No. 111019410
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019410 100 mL
111016646 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019410 Supplier Merck Emd Millipore Supplier No. 79724100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1310-73-2 | 40.00 g/mol

Chemical Identifiers

MDL Number MFCD00003548

Specifications

CAS 1310-73-2
Linear Formula NaOH
Quantity 100 mL
Molecular Weight (g/mol) 40.00
Formula Weight 40.00
Grade HPCE
Concentration or Composition (by Analyte or Components) 0.9-1.1 M
Vapor Pressure 3 mmHg (37°C)
Chemical Name or Material Sodium Hydroxide