missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide solution, Volumetric, Reag. Ph. Eur., 0.1 M NaOH (0.1N), Honeywell Fluka™

Catalog No. p-7072169
Change view
Click to view available options
Quantity:
5 L
10 L
6 x 1 L
Packaging:
Plastic Bottle
VOLPAC™ CONTAINER
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6002632 5 L VOLPAC™ CONTAINER
6002631 5 L Plastic Bottle
60046557 6 x 1 L Plastic Bottle
6002629 10 L VOLPAC™ CONTAINER
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6002632 Supplier Honeywell Chemicals Supplier No. 352635LVP
Only null left

Volumetric, Reag. Ph. Eur., 0.1 M NaOH (0.1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Chemical Identifiers

Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Linear Formula NaOH
Concentration 0.1M
UN Number NONH for all modes of transport
Quantity 5 L
Formula Weight 40g/mol
Packaging VOLPAC™ CONTAINER
Vapor Pressure 3 mmHg (37°C)
Chemical Name or Material Sodium hydroxide solution