missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Monohydrate, Honeywell Fluka™

Catalog No. p-7071694
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001399 25 g
6001398 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001399 Supplier Honeywell Chemicals Supplier No. 0196825G
Only null left

TraceSELECT™, for trace analysis,≥99.9995% (metals basis)

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

Chemical Identifiers

CAS 12200-64-5
Molecular Formula H3NaO2
Molecular Weight (g/mol) 58.012
MDL Number MFCD00149229
InChI Key JBJWASZNUJCEKT-UHFFFAOYSA-M
Synonym sodium hydroxide monohydrate, na.ho.h2o, naoh water, water naoh, sodium hydroxide h2o, sodium hydroxide water, water sodium hydroxide, h2o naoh, naoh h2o, sodium,hydroxide,hydrate
PubChem CID 23719242
IUPAC Name sodium;hydroxide;hydrate
SMILES O.[OH-].[Na+]

Specifications

Linear Formula NaOH · H2O
UN Number 1823
Quantity 25 g
Formula Weight 58.01g/mol
Percent Purity ≥99.99% (metals basis)
Grade TraceSELECT
Chemical Name or Material Sodium Hydroxide Monohydrate
Hazard Statement P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H314
EINECSNumber 215-185-5