missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide monohydrate, 99.996% (metals basis)

Catalog No. AA4128114
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4128114 25 g
AA4128106 5 g
AA4128122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4128114 Supplier Thermo Scientific Chemicals Supplier No. 04128114

CAS: 12179-02-1 | H3NaO2 | 58.01 g/mol

Sodium hydroxide monohydrate is used as an acid neutralizer. It is a strong base widely used in the chemical industry to drive chemical reactions. It finds application as a neutralizing agent in the petroleum refining industry, as paint stripper and an analytical reagent in laboratories to measure the concentration of acids by titration. It is involved in soap production, alumina production by Bayer process, paper industry. It plays an important role as a catalyst for the transesterification of methanol and triglycerides in order to prepare biodiesel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12179-02-1
Molecular Formula H3NaO2
Molecular Weight (g/mol) 58.01
MDL Number MFCD00149229
InChI Key JBJWASZNUJCEKT-UHFFFAOYSA-M
Synonym sodium hydroxide monohydrate, na.ho.h2o, naoh water, water naoh, sodium hydroxide h2o, sodium hydroxide water, water sodium hydroxide, h2o naoh, naoh h2o, sodium,hydroxide,hydrate
PubChem CID 23719242
SMILES O.[OH-].[Na+]

Specifications

Color White
Boiling Point 1390°C
Melting Point 318.4°C
Assay Percent Range 99.996% (metals basis)
Density 2.13 g/cm3 at 20°C
Solubility Information Soluble in water and methanol.
Quantity 25 g
IUPAC Name sodium;hydroxide;hydrate
Formula Weight 58.01 (40.00 Anhydrous)
Odor Odorless
Physical Form Crystals
Chemical Name or Material Sodium hydroxide monohydrate
Show More Show Less
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 215-185-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only