missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 50% (w/w), Solution, 48-52%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor S131025GL
Specifications
| Sodium Hydroxide Solution | |
| 2.5 gal. | |
| Reagent | |
| Poly Drum |
| 1310-73-2,7732-18-5 | |
| 48 to 52% | |
| To Pass Test |
Chemical Identifiers
| 1310-73-2 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.