missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 50% (w/w), Solution, 48-52%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S13104LTPL

 View more versions of this product

Catalog No. 18613618


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Hydroxide Solution
4 L
Reagent
Poly Bottle
1310-73-2,7732-18-5
48 to 52%
To Pass Test

Chemical Identifiers

1310-73-2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.