missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 50% (w/w) Aqueous Solution, Purified, Nitrogen Free, For Kjeldahl Nitrogen, Ricca Chemical
Specifications
Specifications
| CAS | 1310-73-2, 7732-18-5 |
| Molecular Formula | HNaO |
| Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
| Quantity | 10 L |
| InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
| SMILES | [OH-].[Na+] |
| IUPAC Name | sodium hydroxide |
| Molecular Weight (g/mol) | 40.00 |
| PubChem CID | 14798 |
| ChEBI | CHEBI:32145 |
| Show More |