missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 2% (w/v), Ricca Chemical
$258.00 - $301.93
Specifications
| Chemical Name or Material | Sodium Hydroxide |
|---|---|
| Color | Colorless |
| Boiling Point | 100°C |
| CAS | 1310-73-2,1310-73-2,7732-18-5 |
| Melting Point | 0.0°C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-213-113
|
Ricca Chemical Company
R7244200500A |
500 mL |
Each for $258.00
|
|
|||||
|
01-213-118
|
Ricca Chemical Company
R72442001A |
1 L |
Each for $301.93
|
|
|||||
Specifications
| Sodium Hydroxide | |
| 100°C | |
| 0.0°C | |
| 98.04%,1.96% | |
| Laboratory | |
| NIST SRM# 723 |
| Colorless | |
| 1310-73-2,1310-73-2,7732-18-5 | |
| 1.96%,98.04% | |
| 2% | |
| 2% (w/v) | |
| Liquid |