missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 2 N Aqueous Solution, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S375200LTBL

 View more versions of this product

Catalog No. 18613582

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Hydroxide Solution
2 N ± 0.03 N
Chemical Solution
1310-73-2,7732-18-5
200 L
Poly Drum

Chemical Identifiers

1310-73-2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.