missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 0.500 N (N/2), Ricca Chemical
Supplier: Ricca Chemical Company 740032
Description
- Tested in an ISO 17025 Accredited Facility
- Test Reports provided with Certified Values and Measurement Uncertainty
- Tight release specifications for low lot-to-lot variability
Specifications
| Sodium Hydroxide | |
| 94 | |
| HNaO | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| Laboratory | |
| Liquid |
| Colorless | |
| 98 | |
| 0.500 N | |
| 1 L | |
| [OH-].[Na+] | |
| 40 g/mol | |
| CHEBI:32145 | |
| Natural Poly Bottle |
Chemical Identifiers
| HNaO | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium;hydroxide |
| 40 g/mol | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |