missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.500 N (N/2), Ricca Chemical

Supplier:  Ricca Chemical Company 740025

 View more versions of this product

Catalog No. 74002-5


Only null left
Explore available promotions

Description

Description

  • Tested in an ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability
Specifications

Specifications

Sodium Hydroxide
94
HNaO
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
Laboratory
Liquid
Colorless
98
0.500 N
10 L
[OH-].[Na+]
40 g/mol
CHEBI:32145
Cubitainer

Chemical Identifiers

HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
40 g/mol
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions