missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 0.0227 N, 1 mL = 1 mg CO2, Ricca Chemical
Specifications
Specifications
| CAS | 1310-73-2, 7732-18-5 |
| CAS Min % | 98.0, 0.17 |
| CAS Max % | 100.0, 0.17 |
| Molecular Formula | HNaO |
| Concentration or Composition Notes | 0.0227Normal |
| Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
| Quantity | 20 L |
| InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
| SMILES | [OH-].[Na+] |
| IUPAC Name | sodium;hydroxide |
| Show More |