missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrosulfite, tech., 85+%

CAS: 7775-14-6 | Na2O4S2 | 174.096 g/mol

$163.67 - $163.67

Chemical Identifiers

CAS 7775-14-6
Molecular Formula Na2O4S2
Molecular Weight (g/mol) 174.096
MDL Number MFCD00011640
InChI Key JVBXVOWTABLYPX-UHFFFAOYSA-L
Synonym sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros
PubChem CID 24489
ChEBI CHEBI:66870
SMILES [O-]S(=O)S(=O)[O-].[Na+].[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA33381A1
View Documents
Thermo Scientific Chemicals
033381A1
1 kg N/A
Only null left
 
Description

Description

Sodium hydrosulfite is used as a reducing agent in aqueous solutions, sulfonating agent , chelating agent and decolorizing agent in organic reactions. It finds application in water treatment, gas purification, cleaning, leather, polymers, photography, and many others. It is involved in chemical enhanced oil recovery to stabilize polyacrylamide polymers against radical degradation in the presence of iron. It plays an important role to determine the iron content in soil chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7775-14-6
174.096
JVBXVOWTABLYPX-UHFFFAOYSA-L
24489
[O-]S(=O)S(=O)[O-].[Na+].[Na+]
Na2O4S2
MFCD00011640
sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros
CHEBI:66870

Specifications

52°C (decomposition)
White
1 kg
Na2O4S2
sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros
JVBXVOWTABLYPX-UHFFFAOYSA-L
174.096
CHEBI:66870
Moisture sensitive
Sodium hydrosulfite
7775-14-6
Powder
>85%
MFCD00011640
Soluble in water. Slightly soluble in alcohol.
[O-]S(=O)S(=O)[O-].[Na+].[Na+]
24489
174.11
White powder
Videos
Safety and Handling

Safety and Handling

P235+P410-P264b-P270-P280-P301+P312-P305+P351+P338-P330-P501c

H302-H319-X

DOTInformation : DOT Class: 4.2; Packing Group: II

EINECSNumber : 231-890-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only