Learn More
Sodium hydrosulfide hydrate, pure, flakes
CAS: 207683-19-0 | HNaS | 56.06 g/mol
Supplier: Thermo Scientific Chemicals 296200025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
| Sodium hydrosulfide hydrate | |
| 207683-19-0 | |
| 2.5 kg | |
| NaSH·xH2O | |
| 03,266 | |
| 14,8587 | |
| Solubility in water: 620g/L (20°C). Other solubilities: soluble in alcohol and ether | |
| [Na+].[SH-] | |
| 56.06 | |
| 56.06 | |
| Pure | |
| Plastic Bottle |
| 55.0°C | |
| Deliquescing Flakes | |
| HNaS | |
| MFCD00149796 | |
| 90°C | |
| sodium hydrosulfide hydrate, hnas.h2o, nahs.h2o, sodium sulfanide hydrate, sodium bisulfide monohydrate, sodium hydrosulphide hydrate, sodium hydro sulfide hydrate, sodium hydrosulfide monohydrate, sodium hydrate sulfanide, sodium hydrosulfide hydrate, cp | |
| HYHCSLBZRBJJCH-UHFFFAOYSA-M | |
| sodium sulfanide | |
| 16211346 | |
| ≥67% min. (NaHS) typically 70 to 72% | |
| Obnoxious |
Chemical Identifiers
| 207683-19-0 | |
| 56.06 | |
| HYHCSLBZRBJJCH-UHFFFAOYSA-M | |
| 16211346 | |
| [Na+].[SH-] |
| HNaS | |
| MFCD00149796 | |
| sodium hydrosulfide hydrate, hnas.h2o, nahs.h2o, sodium sulfanide hydrate, sodium bisulfide monohydrate, sodium hydrosulphide hydrate, sodium hydro sulfide hydrate, sodium hydrosulfide monohydrate, sodium hydrate sulfanide, sodium hydrosulfide hydrate, cp | |
| sodium sulfanide |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Contact with acids liberates toxic gas.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fre
GHS Signal Word: Danger
RUO – Research Use Only