missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen selenite, 96%

Catalog No. AAA1217318
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1217318 50 g
AAA1217330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1217318 Supplier Thermo Scientific Chemicals Supplier No. A1217318

CAS: 7782-82-3 | Na2O3Se | 172.95 g/mol

Sodium hydrogen selenite is used as a culture media additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydrogen selenite is used as a culture media additive.

Solubility
Soluble in water.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 7782-82-3
Molecular Formula Na2O3Se
Molecular Weight (g/mol) 172.95
MDL Number MFCD00003525
InChI Key BVTBRVFYZUCAKH-UHFFFAOYSA-L
Synonym sodium hydrogen selenite, sodium hydroselenite, sodium biselenite, selenious acid, monosodium salt, sodium selenite nahseo3, sodium hydrogenselenite, hydrogen sodium selenite, unii-him8pp9m79, sodium hydrogen trioxoselenite, hydrogen sodium selenium oxide
PubChem CID 23669629
SMILES [Na+].[Na+].[O-][Se]([O-])=O

Specifications

Quantity 50 g
UN Number UN2630
Solubility Information Soluble in water.
IUPAC Name sodium;hydrogen selenite
Formula Weight 150.96
Percent Purity 98%
Sensitivity Hygroscopic
Chemical Name or Material Sodium hydrogen selenite
Hazard Category H300-H331-H373
Hazard Statement GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENITES
EINECSNumber 231-966-3
RTECSNumber VS7500000
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only