missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen phosphate heptahydrate, ACS, 98.0-102.0%

Catalog No. p-4782145
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1159236 500 g
AA11592A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1159236 Supplier Thermo Scientific Chemicals Supplier No. 01159236
Only null left

CAS: 7782-85-6 | H15Na2O11P | 268.06 g/mol

Sodium hydrogen phosphate is widely used in detergents and cleaning agents. Combined with trisodium phosphate, it is employed in the food industry to adjust the pH and in water treatment to prevent calcium scale formation. It is utilized as a saline laxative to clean the bowel before a colonoscopy. It prevents the coagulation of condensed milk. It is also utilized as anti-caking additive in powdered products. It enhances the cook time and used as thickening agent in desserts and puddings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7782-85-6
Molecular Formula H15Na2O11P
Molecular Weight (g/mol) 268.06
MDL Number MFCD00149180
InChI Key LVXHNCUCBXIIPE-UHFFFAOYSA-L
Synonym sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
PubChem CID 6096963
IUPAC Name disodium phosphoric acid heptahydrate
SMILES O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Melting Point 48.1°C-5H2O
Color White
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 98.0-102.0%
Solubility Information Soluble in water and insoluble in ethanol.
Formula Weight 268.07 (141.98 Anhydrous)
Grade ACS
Odor Odorless
Appearance White crystals
Density 1.7 g/cm3 at 20°C
Chemical Name or Material Sodium hydrogen phosphate heptahydrate
Show More Show Less
Hazard Category H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.
EINECSNumber 231-448-7
RTECSNumber WC4600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only