Learn More
Sodium hydrogen phosphate heptahydrate, ACS, 98.0-102.0%
Description
Sodium hydrogen phosphate is widely used in detergents and cleaning agents. Combined with trisodium phosphate, it is employed in the food industry to adjust the pH and in water treatment to prevent calcium scale formation. It is utilized as a saline laxative to clean the bowel before a colonoscopy. It prevents the coagulation of condensed milk. It is also utilized as anti-caking additive in powdered products. It enhances the cook time and used as thickening agent in desserts and puddings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 48.1°C-5H2O |
| Color | White |
| Physical Form | Crystalline |
| Quantity | 2 kg |
| Odor | Odorless |
| Appearance | White crystals |
| Assay Percent Range | 98.0-102.0% |
| Density | 1.7 |
| Solubility Information | Soluble in water and insoluble in ethanol. |
| Formula Weight | 268.07 (141.98 Anhydrous) |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Hazard Statement | In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste. |
| EINECSNumber | 231-448-7 |
| RTECSNumber | WC4600000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only