missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen L-tartrate, anhydrous, 98%

CAS: 526-94-3 | C4H5NaO6 | 172.07 g/mol

Supplier:  Thermo Scientific Chemicals L1246922

 View more versions of this product

Catalog No. AAL1246922


Only null left
Explore available promotions

Description

Description

Sodium hydrogen L-tartrate is used for detecting potassium and in nutrient media. It also used as a food additive and an acidity regulator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydrogen L-tartrate is used for detecting potassium and in nutrient media. It also used as a food additive and an acidity regulator.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Sodium hydrogen L-tartrate
526-94-3
C4H5NaO6
100 g
14,8589
Soluble in water.
+24.5° (c=3 in Water)
sodium;2,3,4-trihydroxy-4-oxobutanoate
23690454
98%
anhydrous
∽243°C (decomposition)
MFCD00065393,MFCD00065393
6099125
sodium bitartrate, monosodium tartrate, sodium hydrogen tartrate, natriumtartrat german, sodium 3-carboxy-2,3-dihydroxypropanoate, monobasic sodium tartrate, monosodium l-+-tartrate, tartaric acid, monosodium salt, natrium hydrogen-2r,3r-tartrat, weinstein
NKAAEMMYHLFEFN-UHFFFAOYNA-M
[Na+].OC(C(O)C([O-])=O)C(O)=O
172.07
172.07

Chemical Identifiers

526-94-3
172.07
NKAAEMMYHLFEFN-UHFFFAOYNA-M
23690454
[Na+].OC(C(O)C([O-])=O)C(O)=O
C4H5NaO6
MFCD00065393,MFCD00065393
sodium bitartrate, monosodium tartrate, sodium hydrogen tartrate, natriumtartrat german, sodium 3-carboxy-2,3-dihydroxypropanoate, monobasic sodium tartrate, monosodium l-+-tartrate, tartaric acid, monosodium salt, natrium hydrogen-2r,3r-tartrat, weinstein
sodium;2,3,4-trihydroxy-4-oxobutanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-400-9

RTECSNumber : WW8225000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only