missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexanitritocobaltate(III)

Catalog No. AAA1121618
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1121618 50 g
AAA1121630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1121618 Supplier Thermo Scientific Chemicals Supplier No. A1121618
Only null left

CAS: 14649-73-1 | CoN6Na3O12 | 403.933 g/mol

Sodium hexanitritocobaltate(III) is used as primary and secondary intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hexanitritocobaltate(III) is used as primary and secondary intermediate in chemical research.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with acids, reducing agents and amines.

Chemical Identifiers

CAS 14649-73-1
Molecular Formula CoN6Na3O12
Molecular Weight (g/mol) 403.933
MDL Number MFCD00003512
InChI Key ZEBNDJQGEZXBCR-UHFFFAOYSA-H
Synonym cobaltic trisodium hexanitrite, trisodium cobalt 3+ hexanitrite, sodium hexanitritocobaltate 3-, cobalt 3+ sodium nitrite 1:3:6, cobaltate 3-, hexanitrito-, trisodium, cobaltate 3-, hexakis nitrito-o-, trisodium, oc-6-11, na3 co no2 6, cobaltate 3-, hexakis nitrito-kappao-, sodium 1:3 , oc-6-11
PubChem CID 159748
IUPAC Name trisodium;cobalt(3+);hexanitrite
SMILES N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].[Na+].[Na+].[Na+].[Co+3]

Specifications

Melting Point 220°C (decomposition)
Quantity 50 g
Linear Formula Na3Co(NO2)6
UN Number UN1479
Merck Index 14,8603
Solubility Information Soluble in water.
Formula Weight 403.94
Odor Odorless
Chemical Name or Material Sodium hexanitritocobaltate (III)
Hazard Category H272-H315-H317-H319-H334-H335-H350
Hazard Statement GHS H Statement
H272-H334-H351-H335-H315-H319-H317
May intensify fire
oxidizer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P220-P221-P261-P264b-P271-P272-P280-P281-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P342+P311-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
EINECSNumber 237-077-7
RTECSNumber GF9480000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only