missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hexametaphosphate, MilliporeSigma™

Catalog No. MSX05833
Change view
Click to view available options
Quantity:
3 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MSX05833 3 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSX05833 Supplier Technidata Supplier No. SX05833
Only null left

Chemical Identifiers

CAS 10124-56-8
Molecular Formula (NaPO3)6
Synonym Hexasodium Metaphosphate, Hexasodium Salt

Specifications

Physical Form Granules
Quantity 3 kg
Linear Formula (NaPO3)6
Formula Weight 611.8
Packaging Poly Bottle