missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hexametaphosphate, BAKER™, J.T. Baker™

Catalog No. 02004095
Change view
Click to view available options
Quantity:
3 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02004095 3 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02004095 Supplier Avantor Supplier No. V03009

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 10124-56-8
Molecular Formula (NaPO3)6

Specifications

Quantity 3 kg
Formula Weight 611.77
Grade Not Available
Packaging Poly Bottle
Specific Gravity 1L = 1.25kg
Chemical Name or Material Sodium Hexametaphosphate