missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. S039225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S039225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S039225G Supplier TCI America Supplier No. S039225G

Chemical Identifiers

CAS 21324-39-0
Molecular Formula F6NaP
Molecular Weight (g/mol) 167.95
MDL Number MFCD00011122
InChI Key KMADQUOCJBLXRP-UHFFFAOYSA-N
Synonym sodium hexafluorophosphate, sodium hexa-fluoro phosphate, sodium hexafluorophosphate v, napf6, sodium hexaflurophosphate, acmc-1cao2, sodium hexafluoro-, e5-phosphanuide, sodium hexafluoro-$l^ 5-phosphanuide, phosphate 1-, hexafluoro-, sodium 1:1
PubChem CID 5147921
IUPAC Name sodium hexafluoro-λ⁵-phosphanuide
SMILES [Na+].F[P-](F)(F)(F)(F)F

Specifications

Color White
Physical Form Crystalline Powder
Quantity 25 g
UN Number 3260
Formula Weight 167.95
Percent Purity ≥98.0% (T)
Chemical Name or Material Sodium Hexafluorophosphate