Learn More
Sodium hexafluorophosphate, 98%
CAS: 21324-39-0 | F6NaP | 167.95 g/mol
$58.42 - $654.93
Chemical Identifiers
| CAS | 21324-39-0 |
|---|---|
| Molecular Formula | F6NaP |
| Molecular Weight (g/mol) | 167.95 |
| MDL Number | MFCD00011122 |
| InChI Key | KMADQUOCJBLXRP-UHFFFAOYSA-N |
| Synonym | sodium hexafluorophosphate, sodium hexa-fluoro phosphate, sodium hexafluorophosphate v, napf6, sodium hexaflurophosphate, acmc-1cao2, sodium hexafluoro-, e5-phosphanuide, sodium hexafluoro-$l^ 5-phosphanuide, phosphate 1-, hexafluoro-, sodium 1:1 |
| PubChem CID | 5147921 |
| SMILES | [Na+].F[P-](F)(F)(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2089806
|
Thermo Scientific Chemicals
B2089806 |
5 g |
N/A
|
|
|||||
|
AAB2089814
|
Thermo Scientific Chemicals
B2089814 |
25 g |
N/A
|
|
|||||
|
AAB2089822
|
Thermo Scientific Chemicals
B2089822 |
100 g |
N/A
|
|
|||||
Description
Sodium hexafluorophosphate is used as an ion pairing agent for the determination of cobalt(II) in water samples. It is used as an important raw material in the preparation of 2-chloro-1,3-bis(dimethylamino)trimethinium hexafluorophosphate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium hexafluorophosphate is used as an ion pairing agent for the determination of cobalt(II) in water samples. It is used as an important raw material in the preparation of 2-chloro-1,3-bis(dimethylamino)trimethinium hexafluorophosphate.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong acids, bases and oxidizing agents.
Chemical Identifiers
| 21324-39-0 | |
| 167.95 | |
| KMADQUOCJBLXRP-UHFFFAOYSA-N | |
| 5147921 |
| F6NaP | |
| MFCD00011122 | |
| sodium hexafluorophosphate, sodium hexa-fluoro phosphate, sodium hexafluorophosphate v, napf6, sodium hexaflurophosphate, acmc-1cao2, sodium hexafluoro-, e5-phosphanuide, sodium hexafluoro-$l^ 5-phosphanuide, phosphate 1-, hexafluoro-, sodium 1:1 | |
| [Na+].F[P-](F)(F)(F)(F)F |
Specifications
| >200°C | |
| 5 g | |
| MFCD00011122 | |
| sodium hexafluorophosphate, sodium hexa-fluoro phosphate, sodium hexafluorophosphate v, napf6, sodium hexaflurophosphate, acmc-1cao2, sodium hexafluoro-, e5-phosphanuide, sodium hexafluoro-$l^ 5-phosphanuide, phosphate 1-, hexafluoro-, sodium 1:1 | |
| KMADQUOCJBLXRP-UHFFFAOYSA-N | |
| sodium hexafluoro-λâµ-phosphanuide | |
| 5147921 | |
| 98% | |
| Hygroscopic | |
| Sodium hexafluorophosphate |
| 21324-39-0 | |
| F6NaP | |
| UN3260 | |
| Soluble in water. | |
| [Na+].F[P-](F)(F)(F)(F)F | |
| 167.95 | |
| 167.95 | |
| Odorless | |
| 2.37 |
Safety and Handling
GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 244-333-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only