missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexafluorophosphate, 98%

Catalog No. AAB2089806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2089806 5 g
AAB2089814 25 g
AAB2089822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2089806 Supplier Thermo Scientific Chemicals Supplier No. B2089806

CAS: 21324-39-0 | F6NaP | 167.95 g/mol

Sodium hexafluorophosphate is used as an ion pairing agent for the determination of cobalt(II) in water samples. It is used as an important raw material in the preparation of 2-chloro-1,3-bis(dimethylamino)trimethinium hexafluorophosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hexafluorophosphate is used as an ion pairing agent for the determination of cobalt(II) in water samples. It is used as an important raw material in the preparation of 2-chloro-1,3-bis(dimethylamino)trimethinium hexafluorophosphate.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong acids, bases and oxidizing agents.

Chemical Identifiers

CAS 21324-39-0
Molecular Formula F6NaP
Molecular Weight (g/mol) 167.95
MDL Number MFCD00011122
InChI Key KMADQUOCJBLXRP-UHFFFAOYSA-N
Synonym sodium hexafluorophosphate, sodium hexa-fluoro phosphate, sodium hexafluorophosphate v, napf6, sodium hexaflurophosphate, acmc-1cao2, sodium hexafluoro-, e5-phosphanuide, sodium hexafluoro-$l^ 5-phosphanuide, phosphate 1-, hexafluoro-, sodium 1:1
PubChem CID 5147921
SMILES [Na+].F[P-](F)(F)(F)(F)F

Specifications

Melting Point >200°C
Quantity 5 g
UN Number UN3260
Solubility Information Soluble in water.
IUPAC Name sodium hexafluoro-λ⁵-phosphanuide
Formula Weight 167.95
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 2.37
Chemical Name or Material Sodium hexafluorophosphate
Show More Show Less
Hazard Category H302+H312+H332-H314
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 244-333-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only