missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexafluoroarsenate(V), 99%, Thermo Scientific Chemicals

Catalog No. p-7080463
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8851709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA8851709 Supplier Thermo Scientific Chemicals Supplier No. 08851709

Sodium hexafluoroarsenate(V) is generally used for pharmaceutical intermediates and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12005-86-6
Molecular Formula AsF6Na
Molecular Weight (g/mol) 211.902
MDL Number MFCD00014228
InChI Key NFXMAZFYHDSPPB-UHFFFAOYSA-N
Synonym sodium hexafluoroarsenate v, potassium hexafluoro-$l^ 5-arsanuide, sodium hexafluoro-??-arsanuide, sodium hexafluoroarsenate, naasf6
PubChem CID 14205726
IUPAC Name sodium;hexafluoroarsenic(1-)
SMILES F[As-](F)(F)(F)(F)F.[Na+]

Specifications

Physical Form Powder
Quantity 10 g
UN Number UN1557
Solubility Information Soluble in water.
Formula Weight 211.9
Percent Purity 99%
Chemical Name or Material Sodium hexafluoroarsenate (V)
Hazard Category H301+H331
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P310-P304+P340-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
RTECSNumber WB2775000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only